Tisk…

2-Chloro-4-fluorobenzoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A14953.06 A14953.14 A14953.22
A14953.06EA 882 CZK
A14953.06 A14953.14 A14953.22
2-Chloro-4-fluorobenzoic acid ≥98%
2-Chloro-4-fluorobenzoic acid

Purity: 98% CAS No.: 2252-51-9 Molecular Formula: C7H4ClFO2 Formula Weight: 174.56 Melting Point: 183-187° Beilstein Reference: 1946215

Vzorec: C₇H₄ClFO₂
Bod tání: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00010615
CAS číslo: 2252-51-9
EINECS: 218-845-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR