1-Chloro-3-fluorobenzene ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13123.14
A13123.22
A13123.36
A13123.14EA
1550
CZK
A13123.14
A13123.22
A13123.36
1-Chloro-3-fluorobenzene ≥99%
1-Chloro-3-fluorobenzene
Purity: 99%
CAS No.: 625-98-9
Molecular Formula: C6H4ClF
Formula Weight: 130.55
Flash Point: 20°(68°F)
Refractive Index: 1.4940
Boiling Point: 126-128°
Beilstein Reference: 2039303
UN No.: 1993
MDL No.: MFCD00000569
Vzorec:
C₆H₄ClF Bod varu: 126…128 °C Hustota: 1,224 Bod vzplanutí: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000569 CAS číslo: 625-98-9 EINECS: 210-919-0 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...