5-Chloro-2-nitrobenzoic acid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
A12493.06
A12493.14
A12493.22
A12493.06EA
707
CZK
A12493.06
A12493.14
A12493.22
5-Chloro-2-nitrobenzoic acid ≥99%
5-Chloro-2-nitrobenzoic acid
Purity: 99%
CAS No.: 2516-95-2
Molecular Formula: C7H4ClNO4
Formula Weight: 201.57
Odour: Odourless
Melting Point: 136-140°
Beilstein Reference: 1963952
MDL No.: MFCD00007290
Vzorec:
C₇H₄ClNO₄ Bod tání: 136…140 °C Hustota: 1,593 g/cm³ (18 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007290 CAS číslo: 2516-95-2 EINECS: 219-738-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...