Tisk…

2-Chloro-3-nitrobenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

L14954.03 L14954.06
L14954.03EA 1910 CZK
L14954.03 L14954.06
2-Chloro-3-nitrobenzoic acid ≥98%
2-Chloro-3-nitrobenzoic acid

Purity: 98% CAS No.: 3970-35-2 Molecular Formula: C7H4ClNO4 Formula Weight: 201.57 Odour: Odourless Melting Point: 182-186° Beilstein Reference: 645427 MDL No.: MFCD00007069

Vzorec: C₇H₄ClNO₄
Bod tání: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00007069
CAS číslo: 3970-35-2
EINECS: 223-590-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR