4-Amino-3,5-dichlorobenzoic acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
A14982.06
A14982.14
A14982.06EA
1240
CZK
A14982.06
A14982.14
4-Amino-3,5-dichlorobenzoic acid 98%
4-Amino-3,5-dichlorobenzoic acid
Purity: 98%
CAS No.: 56961-25-2
Molecular Formula: C7H5Cl2NO2
Formula Weight: 206.03
Melting Point: ca 290° dec.
Beilstein Reference: 2805751
Vzorec:
C₇H₅Cl₂NO₂ MW: 206,03 g/mol Bod tání: ∼ 290 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00017091 CAS číslo: 56961-25-2 EINECS: 260-468-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...