Tisk…

4-Chloroanthranilic acid 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Amino-4-chlorobenzoic acid

A10201.18 A10201.14 A10201.22 A10201.30
A10201.18EA 2900 CZK
A10201.18 A10201.14 A10201.22 A10201.30
4-Chloroanthranilic acid 98%
4-Chloroanthranilic acid

Purity: 98% CAS No.: 89-77-0 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: ca 232° dec. Beilstein Reference: 743349

Vzorec: C₇H₆ClNO₂
MW: 171,58 g/mol
Bod varu: 330 °C (1013 hPa)
Bod tání: ∼ 232 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00007778
CAS číslo: 89-77-0
EINECS: 201-938-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR