3,4-Dichlorobenzoic acid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
A15873.14
A15873.22
A15873.36
A15873.14EA
941
CZK
A15873.14
A15873.22
A15873.36
3,4-Dichlorobenzoic acid ≥99%
3,4-Dichlorobenzoic acid
Purity: 99%
CAS No.: 51-44-5
Molecular Formula: C7H4Cl2O2
Formula Weight: 191.01
Melting Point: 205-208°
Beilstein Reference: 2044777
Vzorec:
C₇H₄Cl₂O₂ MW: 191,01 g/mol Bod varu: 301 °C (1013 hPa) Bod tání: 205…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002492 CAS číslo: 51-44-5 EINECS: 200-099-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...