Tisk…

2,3-Dichlorobenzoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A10522.09 A10522.18
A10522.09EA 1720 CZK
A10522.09 A10522.18
2,3-Dichlorobenzoic acid ≥98%
2,3-Dichlorobenzoic acid

Purity: 98% CAS No.: 50-45-3 Molecular Formula: C7H4Cl2O2 Formula Weight: 191.01 Melting Point: 166-170° Beilstein Reference: 1946217

Vzorec: C₇H₄Cl₂O₂
Bod tání: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002413
CAS číslo: 50-45-3
EINECS: 200-039-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR