Tisk…

2-Chloro-5-nitrobenzonitrile ≥99%

Dodavatel: Thermo Scientific
Varování

B24287.06 B24287.14 B24287.22
B24287.06EA 917 CZK
B24287.06 B24287.14 B24287.22
2-Chloro-5-nitrobenzonitrile ≥99%
2-Chloro-5-nitrobenzonitrile

Purity: 99% CAS No.: 16588-02-6 Molecular Formula: C7H3ClN2O2 Formula Weight: 182.57 Melting Point: 104-107° Beilstein Reference: 1818642

Vzorec: C₇H₃ClN₂O₂
MW: 182,57 g/mol
Bod varu: 119…122 °C (0,6 mmHg)
Bod tání: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00007292
CAS číslo: 16588-02-6
EINECS: 240-643-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR