2,4-Dichlorobenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A10113.06
A10113.14
A10113.06EA
1270
CZK
A10113.06
A10113.14
2,4-Dichlorobenzonitrile ≥98%
2,4-Dichlorobenzonitrile
Purity: 98%
CAS No.: 6574-98-7
Molecular Formula: C7H3Cl2N
Formula Weight: 172.01
Melting Point: 57-61°
Beilstein Reference: 637738
UN No.: 3439
MDL No.: MFCD00016373
Vzorec:
C₇H₃Cl₂N MW: 172,01 g/mol Bod varu: 261 °C (1013 hPa) Bod tání: 57…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016373 CAS číslo: 6574-98-7 EINECS: 229-493-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...