Tisk…

4-Chlorobenzonitrile ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: p-Chlorobenzonitrile

A14524.0B A14524.18 A14524.30 AB114673.0050
A14524.0BEA 25640 CZK
A14524.0B A14524.18 A14524.30 ABCRAB114673.0050
4-Chlorobenzonitrile ≥99%
4-Chlorobenzonitrile

Purity: 99% CAS No.: 623-03-0 Molecular Formula: C7H4ClN Formula Weight: 137.57 Flash Point: 108°(226°F) Odour: Almond-like Boiling Point: 222-224° Beilstein Reference: 1072122 MDL No.: MFCD00001813

Vzorec: C₇H₄ClN
MW: 137.57
Bod varu: 222…224 °C
Bod tání: 90…94 °C
Hustota: 1.1133
Bod vzplanutí: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001813
CAS číslo: 623-03-0
EINECS: 210-765-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR