5-Chlorosalicylaldehyde ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5-Chloro-2-hydroxybenzaldehyde
A14069.06
A14069.14
A14069.22
A14069.06EA
1460
CZK
A14069.06
A14069.14
A14069.22
5-Chlorosalicylaldehyde ≥98%
5-Chlorosalicylaldehyde
Purity: 98%
CAS No.: 635-93-8
Molecular Formula: C7H5ClO2
Formula Weight: 156.57
Melting Point: 99-103°
Beilstein Reference: 636632
Vzorec:
C₇H₅ClO₂ Bod varu: 105 °C (12 mmHg) Bod tání: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003331 CAS číslo: 635-93-8 EINECS: 211-244-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...