Tisk…

3,5-Dichlorobenzaldehyde ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13325.14 A13325.06
A13325.14EA 8580 CZK
A13325.14 A13325.06
3,5-Dichlorobenzaldehyde ≥97%
3,5-Dichlorobenzaldehyde

Purity: 97% CAS No.: 10203-08-4 Molecular Formula: C7H4Cl2O Formula Weight: 175.01 Melting Point: 60-66° Boiling Point: 235-240° UN No.: 1759 MDL No.: MFCD00003352

Vzorec: C₇H₄Cl₂O
MW: 175.01
Bod varu: 235…240 °C
Bod tání: 60…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00003352
CAS číslo: 10203-08-4
EINECS: 233-499-0
UN: 1759
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR