Tisk…

2,5-Dichlorobenzaldehyde ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A13525.06 A13525.14 A13525.22
A13525.06EA 3100 CZK
A13525.06 A13525.14 A13525.22
2,5-Dichlorobenzaldehyde ≥98%
2,5-Dichlorobenzaldehyde

Purity: 98% CAS No.: 6361-23-5 Molecular Formula: C7H4Cl2O Formula Weight: 175.01 Flash Point: 110°(230°F) Melting Point: 53-58° Boiling Point: 231-233° Beilstein Reference: 1635476 UN No.: 1759 MDL No.: MFCD00156140

Vzorec: C₇H₄Cl₂O
MW: 175,01 g/mol
Bod varu: 231…233 °C
Bod tání: 53…58 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00156140
CAS číslo: 6361-23-5
EINECS: 228-832-1
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR