Tisk…

2,4-Dichlorobenzaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12525.0E A12525.22 A12525.36
A12525.0EEA 14110 CZK
A12525.0E A12525.22 A12525.36
2,4-Dichlorobenzaldehyde ≥98%
2,4-Dichlorobenzaldehyde

Purity: 98% CAS No.: 874-42-0 Molecular Formula: C7H4Cl2O Formula Weight: 175.01 Flash Point: 127°(260°F) Melting Point: 69-73° Boiling Point: 233° Beilstein Reference: 471601 UN No.: 1759 MDL No.: MFCD00003305

Vzorec: C₇H₄Cl₂O
MW: 175.01
Bod varu: 233 °C
Bod tání: 69…73 °C
Hustota: 1.33 g/cm³ (20 °C)
Bod vzplanutí: 127 °C (260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003305
CAS číslo: 874-42-0
EINECS: 212-861-1
UN: 1759
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR