Tisk…

Chlorbenzen, Supelco®

Dodavatel: Merck
Nebezpečí

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

M-Clarity™ quality level = MQ100

08650-5ML-F 08650-25ML-F
08650-5ML-FEA 4440 CZK
08650-5ML-F 08650-25ML-F
Chlorbenzen, Supelco®
Chlorbenzen
Vzorec: C₆H₅Cl
MW: 112.56
Bod varu: 130
Bod tání: –45 °C
Hustota: 1.11 g/cm³ (20 °C)
Bod vzplanutí: 28
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS číslo: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3 III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6ba2f9f60365cc3a976a66389e6ba886d96721c&vid=7933945fd39b21db577f22c9d2753af45a5a1dc2&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR