Chlorbenzen ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Phenylchloride, Benzene Chloride, Monochlorobenzene
B21052.0F
B21052.AP
B21052.0FEA
2430
CZK
B21052.0F
B21052.AP
Chlorbenzen ≥99%
Chlorbenzen
Purity: 99%
CAS No.: 108-90-7
Molecular Formula: C6H5Cl
Formula Weight: 112.56
Flash Point: 28°(82°F)
Refractive Index: 1.5240
Odour: Aromatic
Melting Point: -45°
Boiling Point: 132°
Beilstein Reference: 605632
Merck Reference: 14,2121
UN No.: 1134
MDL No.: MFCD00000530
Vzorec:
C₆H₅Cl Bod varu: 132 °C Bod tání: –45 °C Hustota: 1,106 Bod vzplanutí: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000530 CAS číslo: 108-90-7 EINECS: 203-628-5 UN: 1134 ADR: 3,III REACH: 01-2119432722-45 Merck Index: 14,2121 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...