Tisk…

Chlorbenzen ≥99.5% ACS

Dodavatel: Thermo Fisher Scientific
Varování

36401.AP 36401.K2 36401.K7 36401.M1
36401.APEA 1970 CZK
36401.AP 36401.K2 36401.K7 36401.M1
Chlorbenzen ≥99.5% ACS
Chlorbenzen

Purity: 99.5% CAS No.: 108-90-7 Molecular Formula: C6H5Cl Formula Weight: 112.56 Flash Point: 28°(82°F) Physical Form: Liquid Density: 1.106 Refractive Index: 1.5240 Appearance: Colourless Odour: Aromatic Melting Point: -45° Boiling Point: 132° Beilstein Reference: 605632 Merck Reference: 14,2121 UN No.: 1134 MDL No.: MFCD00000530

Vzorec: C₆H₅Cl
Bod varu: 132 °C
Bod tání: –45 °C
Hustota: 1,106
Bod vzplanutí: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS číslo: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 14,2121
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR