1,3,5-Trichlorobenzene ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
L02132.18
L02132.30
L02132.18EA
948
CZK
L02132.18
L02132.30
1,3,5-Trichlorobenzene ≥98%
1,3,5-Trichlorobenzene
Purity: 98%
CAS No.: 108-70-3
Molecular Formula: C6H3Cl3
Formula Weight: 181.45
Flash Point: 126°(258°F)
Melting Point: 62-65°
Boiling Point: 208°
Beilstein Reference: 1635233
Merck Reference: 14,9632
UN No.: 3077
MDL No.: MFCD00000585
Vzorec:
C₆H₃Cl₃ MW: 181,45 g/mol Bod varu: 208 °C Bod tání: 62…65 °C Hustota: 1,4604 g/cm³ (20 °C) Bod vzplanutí: 126 °C (258 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000585 CAS číslo: 108-70-3 EINECS: 203-608-6 UN: 3077 ADR: 9,III Merck Index: 14,9632 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...