1,2,4,5-Tetrachlorobenzene ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A14318.09
A14318.18
A14318.30
A14318.09EA
876
CZK
A14318.09
A14318.18
A14318.30
1,2,4,5-Tetrachlorobenzene ≥98%
1,2,4,5-Tetrachlorobenzene
Purity: 98%
CAS No.: 95-94-3
Molecular Formula: C6H2Cl4
Formula Weight: 215.89
Flash Point: 155°(311°F)
Odour: Unpleasant
Boiling Point: 245°
Beilstein Reference: 1618315
UN No.: 3077
MDL No.: MFCD00000549
Vzorec:
C₆H₂Cl₄ Bod varu: 245 °C Bod tání: 138…142 °C Hustota: 1,86 Bod vzplanutí: 155 °C (311 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000549 CAS číslo: 95-94-3 EINECS: 202-466-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...