Tisk…

Trimesic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid

A15947.18 A15947.22 A15947.30 A15947.36
A15947.18EA 1790 CZK
A15947.18 A15947.22 A15947.30 A15947.36
Trimesic acid ≥98%
Trimesic acid

Purity: 98% CAS No.: 554-95-0 Molecular Formula: C9H6O6 Formula Weight: 210.14 Flash Point: 328°(622°F) Melting Point: 374-376° Beilstein Reference: 2053080 MDL No.: MFCD00002517

Vzorec: C₉H₆O₆
MW: 210,14 g/mol
Bod tání: 374…376 °C
Bod vzplanutí: 328 °C (622 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002517
CAS číslo: 554-95-0
EINECS: 209-077-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR