Tisk…

Trimellitic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1,2,4-Benzenetricarboxylic acid, Benzene-1,2,4-tricarboxylic acid

L07359.14 L07359.22
L07359.14EA 1310 CZK
L07359.14 L07359.22
Trimellitic acid ≥98%
Trimellitic acid

Purity: 98% CAS No.: 528-44-9 Molecular Formula: C9H6O6 Formula Weight: 210.14 Melting Point: ca 231° dec. Beilstein Reference: 2214815 Merck Reference: 14,9702 MDL No.: MFCD00002470

Vzorec: C₉H₆O₆
MW: 210.14
Bod varu: 107…113 °C (0.1 mmHg)
Bod tání: ∼ 231 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00002470
CAS číslo: 528-44-9
EINECS: 208-432-3
Merck Index: 12,09832

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR