3,5-Dihydroxybenzoic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
alpha-Resorcylic acid
A11664.22
A11664.36
A11664.22EA
1100
CZK
A11664.22
A11664.36
3,5-Dihydroxybenzoic acid ≥97%
3,5-Dihydroxybenzoic acid
Purity: 98%
CAS No.: 99-10-5
Molecular Formula: C7H6O4
Formula Weight: 154.12
Melting Point: ca 237° dec.
Beilstein Reference: 2207864
Vzorec:
C₇H₆O₄ MW: 154.12 Bod tání: ∼237 °C (rozkl.) Bod vzplanutí: >200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002512 CAS číslo: 99-10-5 EINECS: 202-730-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...