Tisk…

Syringic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Hydroxy-3,5-dimethoxybenzoic acid

A11725.14 A11725.22
A11725.14EA 2210 CZK
A11725.14 A11725.22
Syringic acid ≥98%
Syringic acid

Purity: 98+% CAS No.: 530-57-4 Molecular Formula: C9H10O5 Formula Weight: 198.17 Melting Point: 206-208° Boiling Point: 192-193°/14mm MDL No.: MFCD00002552

Vzorec: C₉H₁₀O₅
MW: 198.18
Bod varu: 192…193 °C (14 mmHg)
Bod tání: 206…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00002552
CAS číslo: 530-57-4
EINECS: 208-486-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR