Tisk…

Syringic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Hydroxy-3,5-dimethoxybenzoic acid

132890100 132890250 132891000
ACRO132890100EA 2690 CZK
ACRO132890100 ACRO132890250 ACRO132891000
Syringic acid 97%
Syringic acid
Vzorec: C₉H₁₀O₅
MW: 198.18
Bod varu: 351
Bod tání: 206…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00002552
CAS číslo: 530-57-4
EINECS: 208-486-8

Objednat

Výsledky specifikačního testu

Appearance Beige to grey to light brown Powder
Infrared spectrum Conforms
Melting point 206 to 212 °C
Water ≤0.5 % (K.F.)
Titration with NaOH 96.0 to 104.0 %
NMR (13C-NMR) Consistent with structure
NMR (31P-NMR) Consistent with structure
Elemental Analysis 74.22 to 80.41 % C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR