Tisk…

3,5-Dimethoxybenzoic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A14374.22 A14374.14 A14374.36
A14374.22EA 4500 CZK
A14374.22 A14374.14 A14374.36
3,5-Dimethoxybenzoic acid ≥99%
3,5-Dimethoxybenzoic acid

Purity: 99% CAS No.: 1132-21-4 Molecular Formula: C9H10O4 Formula Weight: 182.18 Odour: Odourless Melting Point: 182-186° Beilstein Reference: 511834 MDL No.: MFCD00002502

Vzorec: C₉H₁₀O₄
MW: 182.18
Bod varu: 341
Bod tání: 182…186 °C
Hustota: 1.423 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002502
CAS číslo: 1132-21-4
EINECS: 214-473-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR