Tisk…

3,4-Dimethoxybenzoic acid ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: Veratric acid

A10167.0B A10167.18 A10167.30
A10167.0BEA 13460 CZK
A10167.0B A10167.18 A10167.30
3,4-Dimethoxybenzoic acid ≥99%
3,4-Dimethoxybenzoic acid

Purity: 99+% CAS No.: 93-07-2 Molecular Formula: C9H10O4 Formula Weight: 182.18 Odour: Odourless Melting Point: 179-183° Beilstein Reference: 518285 Merck Reference: 14,9953 MDL No.: MFCD00002500

Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Bod varu: 303 °C (1013 hPa)
Bod tání: 179…183 °C
Hustota: 1,436 g/cm³ (–100 °C)
Bod vzplanutí: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002500
CAS číslo: 93-07-2
EINECS: 202-215-7
Merck Index: 14,9953

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR