Tisk…

2,4-Dimethoxybenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

A16252.14 A16252.22
A16252.14EA 1270 CZK
A16252.14 A16252.22
2,4-Dimethoxybenzoic acid ≥98%
2,4-Dimethoxybenzoic acid

Purity: 98% CAS No.: 91-52-1 Molecular Formula: C9H10O4 Formula Weight: 182.18 Odour: Odourless Melting Point: 107-111° Beilstein Reference: 2210860 MDL No.: MFCD00002434

Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Bod varu: 313 °C (1013 hPa)
Bod tání: 107…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00002434
CAS číslo: 91-52-1
EINECS: 202-074-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR