p-Anisic acid for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Methoxybenzoic acid, 4-Anisic acid
M-Clarity™ quality level = MQ200
8.05971.0250
8.05971.1000
8.05971.0250EA
1570
CZK
8.05971.0250
8.05971.1000
p-Anisic acid for synthesis, Sigma-Aldrich®
p-Anisic acid
Vzorec:
C₈H₈O₃ MW: 152,15 g/mol Bod varu: 275 °C (1013 hPa) Bod tání: 182…185 °C Hustota: ∼1,385 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002542 CAS číslo: 100-09-4 EINECS: 202-818-5 Merck Index: 13,00669 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...