Tisk…

4-Butoxybenzoic acid ≥98%

Dodavatel: Thermo Fisher Scientific

B21506.14 B21506.22
B21506.14EA 2360 CZK
B21506.14 B21506.22
4-Butoxybenzoic acid ≥98%
4-Butoxybenzoic acid

Purity: 98% CAS No.: 1498-96-0 Molecular Formula: C11H14O3 Formula Weight: 194.23 Melting Point: 146-147° (C-N) MDL No.: MFCD00002546

Vzorec: C₁₁H₁₄O₃
MW: 194.23
Bod varu: 319
Bod tání: 146…147 °C (C-Nematic)
Storage Temperature: Ambient
MDL Number: MFCD00002546
CAS číslo: 1498-96-0
EINECS: 216-104-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR