Tisk…

m-Anisic acid ≥99%

Dodavatel: Thermo Scientific

Synonyms: 3-Anisic acid, 3-Methoxybenzoic acid

A10767.18 A10767.30
A10767.18EA 1090 CZK
A10767.18 A10767.30
m-Anisic acid ≥99%
m-Anisic acid

Purity: 99% CAS No.: 586-38-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless Melting Point: 104-108° Boiling Point: 170-172°/10mm Beilstein Reference: 508838 MDL No.: MFCD00002499

Vzorec: C₈H₈O₃
MW: 152,15 g/mol
Bod varu: 170…172 °C (10 mmHg)
Bod tání: 104…108 °C
Hustota: 1,14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002499
CAS číslo: 586-38-9
EINECS: 209-574-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR