Tisk…

2-Ethoxybenzoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A14758.14 A14758.22 A14758.36
A14758.14EA 1190 CZK
A14758.14 A14758.22 A14758.36
2-Ethoxybenzoic acid ≥98%
2-Ethoxybenzoic acid

Purity: 98% CAS No.: 134-11-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Refractive Index: 1.5410 Melting Point: 19-20° Boiling Point: 174-175°/15mm Beilstein Reference: 2691039 MDL No.: MFCD00002438

Vzorec: C₉H₁₀O₃
Bod varu: 174…175 °C (15 mmHg)
Bod tání: 19…20 °C
Hustota: 1,105
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002438
CAS číslo: 134-11-2
EINECS: 205-130-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR