Tisk…

Kyselina 3,5-dinitrobenzoová ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A16502.0B A16502.30
A16502.0BEA 6250 CZK
A16502.0B A16502.30
Kyselina 3,5-dinitrobenzoová ≥98%
Kyselina 3,5-dinitrobenzoová

Purity: 98+% CAS No.: 99-34-3 Molecular Formula: C7H4N2O6 Formula Weight: 212.12 Melting Point: 204-208° Beilstein Reference: 1914286 Merck Reference: 14,3276 UN No.: 1325 MDL No.: MFCD00007253

Vzorec: C₇H₄N₂O₆
Bod tání: 204…208 °C
Hustota: 1,680
Storage Temperature: Ambient
MDL Number: MFCD00007253
CAS číslo: 99-34-3
EINECS: 202-751-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,3276

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR