Tisk…

2,4-Dinitrobenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

A14536.06 A14536.14
A14536.06EA 948 CZK
A14536.06 A14536.14
2,4-Dinitrobenzoic acid ≥98%
2,4-Dinitrobenzoic acid

Purity: 98% CAS No.: 610-30-0 Molecular Formula: C7H4N2O6 Formula Weight: 212.12 Melting Point: 178-184° Beilstein Reference: 658650

Vzorec: C₇H₄N₂O₆
MW: 212,12 g/mol
Bod tání: 178…184 °C
Hustota: 1,672 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00017006
CAS číslo: 610-30-0
EINECS: 210-219-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR