3-Hydroxy-4-nitrobenzoic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A10724.06
A10724.14
A10724.22
A10724.06EA
749
CZK
A10724.06
A10724.14
A10724.22
3-Hydroxy-4-nitrobenzoic acid ≥98%
3-Hydroxy-4-nitrobenzoic acid
Purity: 98%
CAS No.: 619-14-7
Molecular Formula: C7H5NO5
Formula Weight: 183.12
Melting Point: 232-236°
Beilstein Reference: 2107564
Vzorec:
C₇H₅NO₅ Bod tání: 232…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007110 CAS číslo: 619-14-7 EINECS: 210-580-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...