Tisk…

2,6-Dimethoxy-3-nitrobenzoic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L04366.06 L04366.14
L04366.06EA 1590 CZK
L04366.06 L04366.14
2,6-Dimethoxy-3-nitrobenzoic acid ≥97%
2,6-Dimethoxy-3-nitrobenzoic acid

Purity: 97% CAS No.: 55776-17-5 Molecular Formula: C9H9NO6 Formula Weight: 227.17 Melting Point: 130-132° Beilstein Reference: 2657939

Vzorec: C₉H₉NO₆
Bod tání: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00017324
CAS číslo: 55776-17-5
EINECS: 259-814-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR