3-Nitrobenzoic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
m-Nitrobenzoic acid
A13494.0B
A13494.0I
A13494.30
A13494.0BEA
3330
CZK
A13494.0B
A13494.0I
A13494.30
3-Nitrobenzoic acid ≥99%
3-Nitrobenzoic acid
Purity: 99%
CAS No.: 121-92-6
Molecular Formula: C7H5NO4
Formula Weight: 167.12
Melting Point: 140-143°
Beilstein Reference: 908644
Merck Reference: 14,6589
MDL No.: MFCD00007251
Vzorec:
C₇H₅NO₄ Bod varu: 341 °C (1013 hPa) Bod tání: 140…143 °C Hustota: 1,494 Storage Temperature: Ambient |
MDL Number:
MFCD00007251 CAS číslo: 121-92-6 EINECS: 204-508-5 Merck Index: 14,6589 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...