Tisk…

N-Methylanthranilic acid ≥90%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-(Methylamino)benzoic acid

B20531.14 B20531.22 B20531.36
B20531.14EA 1170 CZK
B20531.14 B20531.22 B20531.36
N-Methylanthranilic acid ≥90%
N-Methylanthranilic acid

Purity: 90+% CAS No.: 119-68-6 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: ca 171° dec. Beilstein Reference: 2208135

Vzorec: C₈H₉NO₂
MW: 151,16 g/mol
Bod varu: 80 °C (0,01 mmHg)
Bod tání: ∼171 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00002424
CAS číslo: 119-68-6
EINECS: 204-343-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR