Tisk…

4-Amino-3-hydroxybenzoic acid 98%

Dodavatel: Thermo Fisher Scientific
Varování

A10207.06 A10207.14 A10207.22
A10207.06EA 2130 CZK
A10207.06 A10207.14 A10207.22
4-Amino-3-hydroxybenzoic acid 98%
4-Amino-3-hydroxybenzoic acid

Purity: 98% CAS No.: 2374-03-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: ca 215° dec. Beilstein Reference: 509859

Vzorec: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00017094
CAS číslo: 2374-03-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR