3-Aminosalicylic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Aminosalicylic Acid, 3-Amino-2-hydroxybenzoic acid, 3-Amino-2-hydroxybenzoic acid
B22478.02
B22478.77
B22478.02EA
6870
CZK
B22478.02
B22478.77
3-Aminosalicylic acid 97%
3-Aminosalicylic acid
Purity: 97%
CAS No.: 570-23-0
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Melting Point: ca 240° dec.
Beilstein Reference: 2090427
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Bod varu: 347 °C (1013 hPa) Bod tání: ∼240 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00010299 CAS číslo: 570-23-0 EINECS: 209-328-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...