4,5-Dimethoxyanthranilic acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Amino-4,5-dimethoxybenzoic acid, 6-Aminoveratric acid
335010050
335010250
335011000
ACRO335010050EA
883
CZK
ACRO335010050
ACRO335010250
ACRO335011000
4,5-Dimethoxyanthranilic acid 98%
4,5-Dimethoxyanthranilic acid
Vzorec:
C₉H₁₁NO₄ MW: 197,19 g/mol Bod varu: 359 °C (1013 hPa) Bod tání: 156…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011671 CAS číslo: 5653-40-7 EINECS: 227-095-3 |
Výsledky specifikačního testu
Appearance | White to beige to grey to brown Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
HPLC | ≥97.5 % |
Titration with NaOH | ≥97.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...