3-Aminobenzoic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A19540.0E
A19540.22
A19540.36
A19540.0EEA
11050
CZK
A19540.0E
A19540.22
A19540.36
3-Aminobenzoic acid 98%
3-Aminobenzoic acid
Purity: 98%
CAS No.: 99-05-8
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 150°(302°F)
Melting Point: 172-179°
Beilstein Reference: 471603
Merck Reference: 14,421
MDL No.: MFCD00007795
Vzorec:
C₇H₇NO₂ Bod varu: 308 °C (1013 hPa) Bod tání: 172…179 °C Hustota: 1,51 Bod vzplanutí: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007795 CAS číslo: 99-05-8 EINECS: 202-724-4 Merck Index: 14,421 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...