Silver benzoate 99%
Dodavatel: Thermo Fisher Scientific
210770100
210770500
ACRO210770100EA
2100
CZK
ACRO210770100
ACRO210770500
Silver benzoate 99%
Silver benzoate
Vzorec:
C₇H₅AgO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00013030 CAS číslo: 532-31-0 EINECS: 208-533-2 Merck Index: 13,01092 |
Výsledky specifikačního testu
Appearance | White to beige to light grey Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Silver (Ag) | 46.4 to 47.8 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...