Tisk…

Benzoan sodný ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: Benzoic acid sodium salt

A15946.36 A15946.0C A15946.0E A15946.30
A15946.36EA 965 CZK
A15946.36 A15946.0C A15946.0E A15946.30
Benzoan sodný ≥99%
Benzoan sodný

Purity: 99% CAS No.: 532-32-1 Molecular Formula: C7H5NaO2 Formula Weight: 144.11 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 300° Solubility: Soluble in water, alcohol Beilstein Reference: 3572467 Merck Reference: 14,8582 MDL No.: MFCD00012463

Vzorec: C₆H₅COONa
MW: 144,11 g/mol
Bod tání: >300 °C
Hustota: 1,44 g/cm³ (20 °C)
Bod vzplanutí: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS číslo: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 14,8582

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR