4-Cyanobenzoic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A15434.22
A15434.06
A15434.14
A15434.22EA
12360
CZK
A15434.22
A15434.06
A15434.14
4-Cyanobenzoic acid ≥98%
4-Cyanobenzoic acid
Purity: 98%
CAS No.: 619-65-8
Molecular Formula: C8H5NO2
Formula Weight: 147.13
Melting Point: ca 221° dec.
Beilstein Reference: 1862865
Vzorec:
C₈H₅NO₂ MW: 147,13 g/mol Bod varu: 313 °C (1013 hPa) Bod tání: ∼221 °C (rozkl.) Hustota: 1,382 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002528 CAS číslo: 619-65-8 EINECS: 210-606-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...