Tisk…

3-Cyanobenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

B23836.06 B23836.14 B23836.22
B23836.06EA 999 CZK
B23836.06 B23836.14 B23836.22
3-Cyanobenzoic acid ≥98%
3-Cyanobenzoic acid

Purity: 98+% CAS No.: 1877-72-1 Molecular Formula: C8H5NO2 Formula Weight: 147.13 Melting Point: 219-224° Beilstein Reference: 1862566

Vzorec: C₈H₅NO₂
MW: 147,13 g/mol
Bod tání: 219…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00002486
CAS číslo: 1877-72-1
EINECS: 217-511-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR