Tisk…

3,4-Dihydroxybenzonitrile ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

A14783.06 A14783.14
A14783.06EA 1730 CZK
A14783.06 A14783.14
3,4-Dihydroxybenzonitrile ≥97%
3,4-Dihydroxybenzonitrile

Purity: 97% CAS No.: 17345-61-8 Molecular Formula: C7H5NO2 Formula Weight: 135.12 Melting Point: 153-156° dec. Beilstein Reference: 2082204 UN No.: 3439 MDL No.: MFCD00016436

Vzorec: C₇H₅NO₂
MW: 135,12 g/mol
Bod varu: 311 °C (1013 hPa)
Bod tání: 153…156 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00016436
CAS číslo: 17345-61-8
EINECS: 241-367-9
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR