Tisk…

Salicylonitrile ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A13606.06 A13606.14 A13606.22 AB117227.0025
A13606.06EA 865 CZK
A13606.06 A13606.14 A13606.22 ABCRAB117227.0025
Salicylonitrile ≥98%
Salicylonitrile

Purity: 98% CAS No.: 611-20-1 Molecular Formula: C7H5NO Formula Weight: 119.12 Melting Point: 92-95° Boiling Point: 149°/14mm MDL No.: MFCD00002145

Vzorec: C₇H₅NO
Bod varu: 149 °C (14 mmHg)
Bod tání: 92…95 °C
Hustota: 1,1052 g/cm³ (99.6 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002145
CAS číslo: 611-20-1
EINECS: 210-259-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR