Tisk…

3-Nitrobenzonitrile ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: m-Nitrobenzonitrile

A15821.09 A15821.14 A15821.30
A15821.09EA 941 CZK
A15821.09 A15821.14 A15821.30
3-Nitrobenzonitrile ≥98%
3-Nitrobenzonitrile

Purity: 98% CAS No.: 619-24-9 Molecular Formula: C7H4N2O2 Formula Weight: 148.12 Odour: Almond-like Melting Point: 114-117° Boiling Point: 165°/21mm Beilstein Reference: 637674 UN No.: 3439 MDL No.: MFCD00007194

Vzorec: C₇H₄N₂O₂
MW: 148,12 g/mol
Bod varu: 165 °C (21 mmHg)
Bod tání: 114…117 °C
Hustota: 0,33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007194
CAS číslo: 619-24-9
EINECS: 210-587-7
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR