Tisk…

2-Nitrobenzonitrile ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: o-Nitrobenzonitrile

A10941.06 A10941.14 A10941.22
A10941.06EA 1070 CZK
A10941.06 A10941.14 A10941.22
2-Nitrobenzonitrile ≥98%
2-Nitrobenzonitrile

Purity: 98+% CAS No.: 612-24-8 Molecular Formula: C7H4N2O2 Formula Weight: 148.12 Odour: Almond-like Melting Point: 107-111° Boiling Point: 165°/16mm Beilstein Reference: 638987 UN No.: 3439 MDL No.: MFCD00007044

Vzorec: C₇H₄N₂O₂
MW: 148,12 g/mol
Bod varu: 165 °C (16 mmHg)
Bod tání: 107…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00007044
CAS číslo: 612-24-8
EINECS: 210-301-0
UN: 3439
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR